Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATTGAAACTATCAACTTTCATG[A/G]AGTTAAGGAAGTTGCGGGAATCAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606029 MIM: 607153 | ||||||||||||||||||||
Literature Links: |
CPSF3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CPSF3 - cleavage and polyadenylation specific factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321833.1 | 891 | Missense Mutation | GAA,GGA | E,G 105 | NP_001308762.1 | |
NM_001321834.1 | 891 | Missense Mutation | GAA,GGA | E,G 105 | NP_001308763.1 | |
NM_001321835.1 | 891 | Intron | NP_001308764.1 | |||
NM_001321836.1 | 891 | Missense Mutation | GAA,GGA | E,G 146 | NP_001308765.1 | |
NM_016207.3 | 891 | Missense Mutation | GAA,GGA | E,G 142 | NP_057291.1 |
ITGB1BP1 - integrin subunit beta 1 binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |