Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGGCCTTATATGATGTCGTACC[A/G]TTGAGTCTTGAAGTCCAGGCAGTTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603817 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASAP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ASAP2 - ArfGAP with SH3 domain, ankyrin repeat and PH domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135191.1 | Intron | NP_001128663.1 | ||||
NM_003887.2 | Intron | NP_003878.1 | ||||
XM_006711898.1 | Intron | XP_006711961.1 | ||||
XM_006711901.3 | Intron | XP_006711964.1 | ||||
XM_006711902.1 | Intron | XP_006711965.1 | ||||
XM_011510403.2 | Intron | XP_011508705.1 | ||||
XM_011510404.2 | Intron | XP_011508706.1 | ||||
XM_011510405.2 | Intron | XP_011508707.1 | ||||
XM_011510406.2 | Intron | XP_011508708.1 | ||||
XM_011510407.2 | Intron | XP_011508709.1 | ||||
XM_011510408.2 | Intron | XP_011508710.1 | ||||
XM_011510409.1 | Intron | XP_011508711.1 |
Set Membership: |
HapMap |