Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACATTTTATTGTCCATGTTCCAGG[C/G]TTCATATGCCCAGCAAAGAAATAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600389 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MEP1B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MEP1B - meprin A subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308171.1 | Intron | NP_001295100.1 | ||||
NM_005925.2 | Intron | NP_005916.2 | ||||
XM_011526013.2 | Intron | XP_011524315.1 | ||||
XM_011526014.2 | Intron | XP_011524316.1 |
Set Membership: |
HapMap |