Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCGTAGGATGATGCATCTTTTAC[C/T]CAACCAATGGGCCAGAGTGAGCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC11A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
SEC11A - SEC11 homolog A, signal peptidase complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271918.1 | Intron | NP_001258847.1 | ||||
NM_001271919.1 | Intron | NP_001258848.1 | ||||
NM_001271920.1 | Intron | NP_001258849.1 | ||||
NM_001271921.1 | Intron | NP_001258850.1 | ||||
NM_001271922.1 | Intron | NP_001258851.1 | ||||
NM_014300.3 | Intron | NP_055115.1 |
Set Membership: |
HapMap |