Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCTGTAGCAGCCGCTGGAGCCG[C/T]GCGCCCTCCCGCAGGCGGCTGAGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606839 MIM: 182099 | ||||||||||||||||||||
Literature Links: |
CDHR5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDHR5 - cadherin related family member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCT - secretin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021920.3 | 225 | Silent Mutation | GCA,GCG | A,A 44 | NP_068739.1 | |
XM_017018109.1 | 225 | Silent Mutation | GCA,GCG | A,A 42 | XP_016873598.1 |