Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGCAGAGGGCCAACAGCCTGAA[A/C]GCCATTCTTAGTTAATCAGGGCAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616160 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHRS7B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DHRS7B - dehydrogenase/reductase 7B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM11 - transmembrane protein 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003876.2 | Intron | NP_003867.1 |