Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCATTCTATATACTATTTAATTCA[C/T]AGACAGAGACTGGAAAAGGAAAAGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603882 MIM: 606144 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAG2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BAG2 - BCL2 associated athanogene 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB23 - RAB23, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278666.1 | 3407 | UTR 3 | NP_001265595.1 | |||
NM_001278667.1 | 3407 | UTR 3 | NP_001265596.1 | |||
NM_001278668.1 | 3407 | UTR 3 | NP_001265597.1 | |||
NM_016277.4 | 3407 | UTR 3 | NP_057361.3 | |||
NM_183227.2 | 3407 | UTR 3 | NP_899050.1 |
Set Membership: |
HapMap |