Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAAGAACATCAAAACAAAATTT[C/T]CCTGACACTGGAAGCTTCTTATATG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614577 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BMP2K PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BMP2K - BMP2 inducible kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017593.3 | Intron | NP_060063.2 | ||||
NM_198892.1 | Intron | NP_942595.1 | ||||
XM_005263117.1 | Intron | XP_005263174.1 | ||||
XM_011532102.1 | Intron | XP_011530404.1 | ||||
XM_017008381.1 | Intron | XP_016863870.1 | ||||
XM_017008382.1 | Intron | XP_016863871.1 |
PAQR3 - progestin and adipoQ receptor family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040202.1 | Intron | NP_001035292.1 | ||||
XM_005262769.4 | Intron | XP_005262826.1 | ||||
XM_017007785.1 | Intron | XP_016863274.1 |
Set Membership: |
HapMap |