Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGAATTGCTATATTTAGAAAGAA[C/T]ATGAAACAGAATATCAGCAATGTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRIQ3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LRRIQ3 - leucine rich repeats and IQ motif containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105659.1 | 1568 | UTR 3 | NP_001099129.1 | |||
NM_001322315.1 | 1568 | UTR 3 | NP_001309244.1 | |||
XM_011540658.1 | 1568 | UTR 3 | XP_011538960.1 | |||
XM_011540659.1 | 1568 | UTR 3 | XP_011538961.1 | |||
XM_011540660.1 | 1568 | UTR 3 | XP_011538962.1 | |||
XM_011540662.2 | 1568 | UTR 3 | XP_011538964.1 | |||
XM_011540663.2 | 1568 | Intron | XP_011538965.1 | |||
XM_017000286.1 | 1568 | Intron | XP_016855775.1 | |||
XM_017000287.1 | 1568 | Intron | XP_016855776.1 | |||
XM_017000288.1 | 1568 | Intron | XP_016855777.1 |