Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTCCCCAGAGTTAAGACAGTCAC[A/G]TACTCCCCACAAATGTCTCTCCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
35 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615394 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NSUN4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NSUN4 - NOP2/Sun RNA methyltransferase family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256127.1 | Intron | NP_001243056.1 | ||||
NM_001256128.1 | Intron | NP_001243057.1 | ||||
NM_199044.3 | Intron | NP_950245.2 |
Set Membership: |
HapMap |