Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGTTATTGGCACAAGTAGCCTCA[G/T]TTCTCAATCCAGATAATTAACTGTG
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||
Literature Links: |
C6orf163 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
C6orf163 - chromosome 6 open reading frame 163 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010868.2 | 144 | Intron | NP_001010868.2 | |||
XM_005248673.3 | 144 | UTR 5 | XP_005248730.1 | |||
XM_006715370.3 | 144 | UTR 5 | XP_006715433.1 | |||
XM_006715372.2 | 144 | Intron | XP_006715435.1 | |||
XM_011535539.1 | 144 | UTR 5 | XP_011533841.1 | |||
XM_017010367.1 | 144 | Intron | XP_016865856.1 |
SMIM8 - small integral membrane protein 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042493.2 | 144 | Intron | NP_001035958.1 | |||
NM_001287445.1 | 144 | Intron | NP_001274374.1 | |||
NM_020425.5 | 144 | Intron | NP_065158.3 |