Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTCATATCAAGAAGTGAAATTCA[C/T]CGGGCAAGCCCCAGCTGGCTCCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRAMD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GRAMD3 - GRAM domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146319.1 | Intron | NP_001139791.1 | ||||
NM_001146320.1 | Intron | NP_001139792.1 | ||||
NM_001146321.1 | Intron | NP_001139793.1 | ||||
NM_001146322.1 | Intron | NP_001139794.1 | ||||
NM_023927.2 | Intron | NP_076416.2 | ||||
XM_005272057.3 | Intron | XP_005272114.1 | ||||
XM_005272058.3 | Intron | XP_005272115.1 | ||||
XM_005272059.4 | Intron | XP_005272116.1 | ||||
XM_005272060.3 | Intron | XP_005272117.1 | ||||
XM_005272061.3 | Intron | XP_005272118.1 | ||||
XM_005272062.3 | Intron | XP_005272119.1 | ||||
XM_005272063.3 | Intron | XP_005272120.1 | ||||
XM_005272064.3 | Intron | XP_005272121.1 | ||||
XM_011543591.2 | Intron | XP_011541893.1 | ||||
XM_011543592.2 | Intron | XP_011541894.1 | ||||
XM_011543593.2 | Intron | XP_011541895.1 | ||||
XM_011543594.2 | Intron | XP_011541896.1 | ||||
XM_011543595.2 | Intron | XP_011541897.1 | ||||
XM_017009781.1 | Intron | XP_016865270.1 | ||||
XM_017009782.1 | Intron | XP_016865271.1 | ||||
XM_017009783.1 | Intron | XP_016865272.1 | ||||
XM_017009784.1 | Intron | XP_016865273.1 | ||||
XM_017009785.1 | Intron | XP_016865274.1 |
LOC105369178 - uncharacterized LOC105369178 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |