Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGACGAAGAGCGCCGGGCCGCGAC[C/T]AGGAGCCCACCCGCGCCCTCCGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107450 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFNAR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IFNAR1 - interferon alpha and beta receptor subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000629.2 | 120 | Intron | NP_000620.2 | |||
XM_005260964.2 | 120 | UTR 5 | XP_005261021.1 | |||
XM_011529552.1 | 120 | Intron | XP_011527854.1 |