Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCGCAGCCAAACCTCATCCCCTT[C/T]GGCTAGCTTCAGCACAGCATGATTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612045 | ||||||||||||||||||||
Literature Links: |
C1QTNF3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1QTNF3 - C1q and tumor necrosis factor related protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030945.3 | 1000 | Missense Mutation | AAA,GAA | K,E 214 | NP_112207.1 | |
NM_181435.5 | 1000 | Missense Mutation | AAA,GAA | K,E 287 | NP_852100.3 |
C1QTNF3-AMACR - C1QTNF3-AMACR readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |