Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGAGAAACAGTAACTCTATGGCA[A/G]GTTTGATTTATATAAAGAGGGTATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603525 | ||||||||||||||||||||
Literature Links: |
NMI PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NMI - N-myc and STAT interactor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004688.2 | 1090 | Silent Mutation | ACC,ACT | T,T 229 | NP_004679.2 | |
XM_005246941.2 | 1090 | Silent Mutation | ACC,ACT | T,T 229 | XP_005246998.1 | |
XM_017005247.1 | 1090 | Silent Mutation | ACC,ACT | T,T 229 | XP_016860736.1 |
RBM43 - RNA binding motif protein 43 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |