Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAAGGAGAAGTTGAGACTCAACC[A/G]GGTAGTTTTTCTTTAATTGTCTTCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601437 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCGRT PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FCGRT - Fc fragment of IgG receptor and transporter | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136019.2 | Intron | NP_001129491.1 | ||||
NM_004107.4 | Intron | NP_004098.1 | ||||
XM_005258657.1 | Intron | XP_005258714.1 | ||||
XM_011526627.2 | Intron | XP_011524929.1 |
Set Membership: |
HapMap |