Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGCCCTTAGGAATTGAGTTTTGA[A/G]GGATGAGTTTAACAGGCTCTGAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 159440 MIM: 602413 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MPZ PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MPZ - myelin protein zero | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SDHC - succinate dehydrogenase complex subunit C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035511.1 | Intron | NP_001030588.1 | ||||
NM_001035512.1 | Intron | NP_001030589.1 | ||||
NM_001035513.1 | Intron | NP_001030590.1 | ||||
NM_001278172.1 | Intron | NP_001265101.1 | ||||
NM_003001.3 | Intron | NP_002992.1 |
Set Membership: |
HapMap |