Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCAAAGTCCACCAGAATGTTAGC[A/G]GATGTGTCCTTGCAGGCCAAGCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
65 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR6N1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OR6N1 - olfactory receptor family 6 subfamily N member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005185.1 | 579 | Intron | NP_001005185.1 | |||
XM_017000325.1 | 579 | Intron | XP_016855814.1 | |||
XM_017000326.1 | 579 | Intron | XP_016855815.1 | |||
XM_017000327.1 | 579 | Intron | XP_016855816.1 |
OR6N2 - olfactory receptor family 6 subfamily N member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005278.1 | 579 | Silent Mutation | TCC,TCT | S,S 193 | NP_001005278.1 |
Set Membership: |
HapMap |