Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGATGATCTAGAGCCTGAATGC[A/G]TCTCAGGGAAGACTTTCTTAAGTAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611042 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LELP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LELP1 - late cornified envelope like proline rich 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010857.2 | Intron | NP_001010857.1 |
SPRR2G - small proline rich protein 2G | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |