Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATGGATATCCGGCTGGAAGCATC[C/T]CCTACCCGCTGGGAGAGTGGGTCTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
27 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114210 MIM: 176991 MIM: 114110 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
S100A4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
S100A4 - S100 calcium binding protein A4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
S100A5 - S100 calcium binding protein A5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
S100A6 - S100 calcium binding protein A6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014624.3 | Intron | NP_055439.1 | ||||
XM_017002033.1 | Intron | XP_016857522.1 |
Set Membership: |
HapMap |