Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACCCTGTGCTCCCACAGACTGCA[A/G]CGTGGGGGACCCTGGGCACCCTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
36 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603319 MIM: 606920 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANXA9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANXA9 - annexin A9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003568.2 | 271 | Missense Mutation | ACG,GCG | T,A 28 | NP_003559.2 | |
XM_011510058.2 | 271 | Missense Mutation | ACG,GCG | T,A 28 | XP_011508360.1 |
CERS2 - ceramide synthase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |