Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAAAGCATTGAGGACTTGCACAC[C/G]TGCCGTCTCTTAATTAAACAGGACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610276 | ||||||||||||||||||||
Literature Links: |
PIGX PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PIGX - phosphatidylinositol glycan anchor biosynthesis class X | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166304.1 | 387 | Silent Mutation | ACC,ACG | T,T 76 | NP_001159776.1 | |
NM_017861.3 | 387 | Silent Mutation | ACC,ACG | T,T 76 | NP_060331.3 |