Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGCTAGGCTGGCTTCCACCTTGC[C/T]TGCTTGCCCCCTGTATACCTCCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611968 MIM: 176894 | ||||||||||||||||||||
Literature Links: |
CSTF2T PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CSTF2T - cleavage stimulation factor subunit 2, tau variant | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015235.2 | 1704 | Missense Mutation | AAG,AGG | K,R 553 | NP_056050.1 |
PRKG1 - protein kinase, cGMP-dependent, type I | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098512.2 | 1704 | Intron | NP_001091982.1 | |||
NM_006258.3 | 1704 | Intron | NP_006249.1 | |||
XM_011539952.2 | 1704 | Intron | XP_011538254.1 | |||
XM_017016412.1 | 1704 | Intron | XP_016871901.1 | |||
XM_017016413.1 | 1704 | Intron | XP_016871902.1 |