Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGAGGTCACTTCTGAGAATAATA[C/T]ATCAAAAAGGGTACTGGTTAGAGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614571 MIM: 608667 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5orf42 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C5orf42 - chromosome 5 open reading frame 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023073.3 | Intron | NP_075561.3 | ||||
XM_005248345.3 | Intron | XP_005248402.1 | ||||
XM_005248346.3 | Intron | XP_005248403.1 | ||||
XM_005248347.3 | Intron | XP_005248404.1 | ||||
XM_005248349.3 | Intron | XP_005248406.1 | ||||
XM_005248350.3 | Intron | XP_005248407.1 | ||||
XM_006714491.3 | Intron | XP_006714554.1 | ||||
XM_011514085.2 | Intron | XP_011512387.1 | ||||
XM_011514086.2 | Intron | XP_011512388.1 | ||||
XM_011514087.2 | Intron | XP_011512389.1 | ||||
XM_011514088.2 | Intron | XP_011512390.1 | ||||
XM_011514089.2 | Intron | XP_011512391.1 | ||||
XM_011514090.2 | Intron | XP_011512392.1 | ||||
XM_011514092.2 | Intron | XP_011512394.1 | ||||
XM_011514094.2 | Intron | XP_011512396.1 | ||||
XM_017009760.1 | Intron | XP_016865249.1 | ||||
XM_017009761.1 | Intron | XP_016865250.1 | ||||
XM_017009762.1 | Intron | XP_016865251.1 | ||||
XM_017009763.1 | Intron | XP_016865252.1 | ||||
XM_017009764.1 | Intron | XP_016865253.1 | ||||
XM_017009765.1 | Intron | XP_016865254.1 | ||||
XM_017009766.1 | Intron | XP_016865255.1 |
NIPBL - NIPBL, cohesin loading factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015384.4 | Intron | NP_056199.2 | ||||
NM_133433.3 | Intron | NP_597677.2 | ||||
XM_005248280.3 | Intron | XP_005248337.1 | ||||
XM_005248282.4 | Intron | XP_005248339.3 | ||||
XM_006714467.2 | Intron | XP_006714530.1 | ||||
XM_006714468.2 | Intron | XP_006714531.1 | ||||
XM_011514015.1 | Intron | XP_011512317.1 | ||||
XM_017009329.1 | Intron | XP_016864818.1 | ||||
XM_017009330.1 | Intron | XP_016864819.1 | ||||
XM_017009331.1 | Intron | XP_016864820.1 |
Set Membership: |
HapMap |