Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTGTTCTGGTTCTCTTGTGGCC[A/G]CCGTCGCTGTCCGGCTGCCTTGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607826 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHCYL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AHCYL1 - adenosylhomocysteinase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242673.1 | 87 | UTR 5 | NP_001229602.1 | |||
NM_001242674.1 | 87 | Intron | NP_001229603.1 | |||
NM_001242675.1 | 87 | Intron | NP_001229604.1 | |||
NM_001242676.1 | 87 | Intron | NP_001229605.1 | |||
NM_006621.5 | 87 | UTR 5 | NP_006612.2 | |||
XM_011540535.1 | 87 | UTR 5 | XP_011538837.1 |