Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGCTTTCTCTGAAGGTGCCTGCA[A/G]TGATATTTAGGCATTGGCCTGTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP5SL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP5SL - ATP5S like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167867.1 | 542 | Intron | NP_001161339.1 | |||
NM_001167868.1 | 542 | Intron | NP_001161340.1 | |||
NM_001167869.1 | 542 | Intron | NP_001161341.1 | |||
NM_001167870.1 | 542 | Intron | NP_001161342.1 | |||
NM_001167871.1 | 542 | Intron | NP_001161343.1 | |||
NM_001320838.1 | 542 | Intron | NP_001307767.1 | |||
NM_001320839.1 | 542 | Intron | NP_001307768.1 | |||
NM_001320840.1 | 542 | Intron | NP_001307769.1 | |||
NM_001320841.1 | 542 | Intron | NP_001307770.1 | |||
NM_001320844.1 | 542 | Intron | NP_001307773.1 | |||
NM_018035.2 | 542 | Intron | NP_060505.2 | |||
XM_011527065.2 | 542 | Intron | XP_011525367.1 |
ERICH4 - glutamate rich 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130514.1 | 542 | UTR 3 | NP_001123986.1 | |||
XM_017026119.1 | 542 | UTR 3 | XP_016881608.1 |
PCAT19 - prostate cancer associated transcript 19 (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |