Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTCAGTTCCCGCATGGGGGCGA[G/T]GCAGGTACATGGGCCTTGCGGGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612665 | ||||||||||||||||||||
Literature Links: |
TEX101 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TEX101 - testis expressed 101 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130011.1 | 572 | Intron | NP_001123483.1 | |||
NM_031451.4 | 572 | Missense Mutation | AGG,ATG | R,M 4 | NP_113639.4 | |
XM_005259303.3 | 572 | Intron | XP_005259360.2 |