Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAAGCTCTCTCCTTCATGTGGAG[C/T]GTGTACCAAGTTACAGTTCCTGCGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601703 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPM1N PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PPM1N - protein phosphatase, Mg2+/Mn2+ dependent 1N (putative) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VASP - vasodilator-stimulated phosphoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003370.3 | Intron | NP_003361.1 | ||||
XM_005259199.1 | Intron | XP_005259256.1 | ||||
XM_005259200.1 | Intron | XP_005259257.1 | ||||
XM_017027200.1 | Intron | XP_016882689.1 |
Set Membership: |
HapMap |