Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCTGCAGCGACAACATTGACTT[C/T]GGCCGCCTCTTCGGCCGGGAGTTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 164820 MIM: 601906 | ||||||||||||||||||||
Literature Links: |
WNT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WNT1 - Wnt family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005430.3 | 726 | Silent Mutation | TTC,TTT | F,F 176 | NP_005421.1 |
WNT10B - Wnt family member 10B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |