Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACATTTTCTAAGCTCCTTCCAAA[C/T]GAATGCATCAATTTTTAATCCAGGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105376204 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105376204 - uncharacterized LOC105376204 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF462 - zinc finger protein 462 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021224.4 | Intron | NP_067047.4 | ||||
XM_006717209.3 | Intron | XP_006717272.1 | ||||
XM_006717211.3 | Intron | XP_006717274.1 | ||||
XM_006717212.3 | Intron | XP_006717275.1 | ||||
XM_006717215.3 | Intron | XP_006717278.1 | ||||
XM_006717216.3 | Intron | XP_006717279.1 | ||||
XM_006717218.3 | Intron | XP_006717281.1 | ||||
XM_011518892.2 | Intron | XP_011517194.1 | ||||
XM_017014996.1 | Intron | XP_016870485.1 | ||||
XM_017014997.1 | Intron | XP_016870486.1 | ||||
XM_017014998.1 | Intron | XP_016870487.1 | ||||
XM_017014999.1 | Intron | XP_016870488.1 |
Set Membership: |
HapMap |