Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCGCTCCCCTTTGCTGAAGAGT[C/T]GGGAGCACAGGAGATCCCAGCTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605584 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHX38 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
DHX38 - DEAH-box helicase 38 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PMFBP1 - polyamine modulated factor 1 binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160213.1 | Intron | NP_001153685.1 | ||||
NM_031293.2 | Intron | NP_112583.2 | ||||
XM_011523357.2 | Intron | XP_011521659.1 | ||||
XM_011523358.2 | Intron | XP_011521660.1 | ||||
XM_011523360.2 | Intron | XP_011521662.1 | ||||
XM_011523361.2 | Intron | XP_011521663.1 | ||||
XM_011523362.2 | Intron | XP_011521664.1 |