Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAGTCGGTGTCTGCTGCGTTCAC[A/G]GCAGGATTCGGTTAGGTGAGTGTGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616434 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IST1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IST1 - IST1, ESCRT-III associated factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270975.1 | 82 | UTR 5 | NP_001257904.1 | |||
NM_001270976.1 | 82 | Intron | NP_001257905.1 | |||
NM_001270977.1 | 82 | UTR 5 | NP_001257906.1 | |||
NM_001270978.1 | 82 | UTR 5 | NP_001257907.1 | |||
NM_001270979.1 | 82 | UTR 5 | NP_001257908.1 | |||
NM_014761.3 | 82 | UTR 5 | NP_055576.2 |
ZNF821 - zinc finger protein 821 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |