Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300172 MIM: 300241 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASK PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CASK - calcium/calmodulin dependent serine protein kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126054.2 | Intron | NP_001119526.1 | ||||
NM_001126055.2 | Intron | NP_001119527.1 | ||||
NM_003688.3 | Intron | NP_003679.2 | ||||
XM_005272686.4 | Intron | XP_005272743.1 | ||||
XM_006724566.3 | Intron | XP_006724629.1 | ||||
XM_011543993.2 | Intron | XP_011542295.1 | ||||
XM_011543994.2 | Intron | XP_011542296.1 | ||||
XM_011543995.2 | Intron | XP_011542297.1 | ||||
XM_011543996.2 | Intron | XP_011542298.1 | ||||
XM_011543997.2 | Intron | XP_011542299.1 |
GPR34 - G protein-coupled receptor 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001097579.1 | Intron | NP_001091048.1 | ||||
NM_005300.3 | Intron | NP_005291.1 | ||||
XM_005272597.3 | Intron | XP_005272654.1 |
Set Membership: |
HapMap |