Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACCAGTGAAAAAGCAATCACTTA[C/T]GATCCTGGGCTTTGAAAAATCACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613777 MIM: 609063 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXRED2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FOXRED2 - FAD dependent oxidoreductase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102371.1 | 3007 | UTR 3 | NP_001095841.1 | |||
NM_024955.5 | 3007 | UTR 3 | NP_079231.4 | |||
XM_017028941.1 | 3007 | UTR 3 | XP_016884430.1 | |||
XM_017028942.1 | 3007 | UTR 3 | XP_016884431.1 |
TXN2 - thioredoxin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |