Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCCTGTCCTGTCAGTTCGGGGTC[C/T]TTGGCCTCATTGTAGAATGGAAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613534 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FAN1 - FANCD2/FANCI-associated nuclease 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146094.1 | Intron | NP_001139566.1 | ||||
NM_001146095.1 | Intron | NP_001139567.1 | ||||
NM_001146096.1 | Intron | NP_001139568.1 | ||||
NM_014967.4 | Intron | NP_055782.3 | ||||
XM_005254232.4 | Intron | XP_005254289.1 | ||||
XM_005254234.4 | Intron | XP_005254291.1 | ||||
XM_005254235.3 | Intron | XP_005254292.1 | ||||
XM_011521370.2 | Intron | XP_011519672.1 | ||||
XM_011521371.1 | Intron | XP_011519673.1 | ||||
XM_011521372.2 | Intron | XP_011519674.1 | ||||
XM_017022012.1 | Intron | XP_016877501.1 | ||||
XM_017022013.1 | Intron | XP_016877502.1 |
MTMR10 - myotubularin related protein 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017762.2 | Intron | NP_060232.2 | ||||
XM_005254508.2 | Intron | XP_005254565.1 | ||||
XM_011521737.2 | Intron | XP_011520039.1 | ||||
XM_011521738.2 | Intron | XP_011520040.1 | ||||
XM_017022384.1 | Intron | XP_016877873.1 |
Set Membership: |
HapMap |