Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601129 | ||||||||||||||||||||||||||||||||
Literature Links: |
CYP2C8 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU) - Not Available | |||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | |||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CYP2C8 - cytochrome P450 family 2 subfamily C member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000770.3 | 1477 | Frame Shift Delete | GTG,GTTGAT | V,D 461V | NP_000761.3 | |
NM_001198853.1 | 1477 | Frame Shift Delete | GTG,GTTGAT | V,D 391V | NP_001185782.1 | |
NM_001198854.1 | 1477 | Frame Shift Delete | GTG,GTTGAT | V,D 359V | NP_001185783.1 | |
NM_001198855.1 | 1477 | Frame Shift Delete | GTG,GTTGAT | V,D 391V | NP_001185784.1 |
Set Membership: |
DME Validated Inventoried |