Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGACTTAATTAGCTGCTTTATCA[C/T]TTCACTGCTGTCTGAAGTACACAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604515 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLNK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BLNK - B-cell linker | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114094.1 | 2601 | Intron | NP_001107566.1 | |||
NM_001258440.1 | 2601 | Intron | NP_001245369.1 | |||
NM_001258441.1 | 2601 | Intron | NP_001245370.1 | |||
NM_001258442.1 | 2601 | Intron | NP_001245371.1 | |||
NM_013314.3 | 2601 | Intron | NP_037446.1 | |||
XM_011539728.2 | 2601 | UTR 3 | XP_011538030.1 | |||
XM_017016159.1 | 2601 | UTR 3 | XP_016871648.1 |
Set Membership: |
HapMap |