Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCTGGCCAAGTGAGATTAAGGCA[A/G]TGTGTTTCATGACAAATGAAATTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCGF5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PCGF5 - polycomb group ring finger 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256549.1 | Intron | NP_001243478.1 | ||||
NM_001257101.1 | Intron | NP_001244030.1 | ||||
NM_032373.4 | Intron | NP_115749.2 | ||||
XM_006718030.1 | Intron | XP_006718093.1 | ||||
XM_011540271.2 | Intron | XP_011538573.1 | ||||
XM_011540272.2 | Intron | XP_011538574.1 | ||||
XM_017016776.1 | Intron | XP_016872265.1 | ||||
XM_017016777.1 | Intron | XP_016872266.1 | ||||
XM_017016778.1 | Intron | XP_016872267.1 |
Set Membership: |
HapMap |