Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTTCTCATCTTCCAGACTGAACA[C/T]GCTTAGTAATAATCAAAACATTCTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605651 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXW11 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FBXW11 - F-box and WD repeat domain containing 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012300.2 | Intron | NP_036432.2 | ||||
NM_033644.2 | Intron | NP_387448.2 | ||||
NM_033645.2 | Intron | NP_387449.2 | ||||
XM_005265855.4 | Intron | XP_005265912.1 | ||||
XM_005265856.2 | Intron | XP_005265913.1 | ||||
XM_005265857.2 | Intron | XP_005265914.1 | ||||
XM_005265858.2 | Intron | XP_005265915.1 | ||||
XM_017009279.1 | Intron | XP_016864768.1 | ||||
XM_017009280.1 | Intron | XP_016864769.1 | ||||
XM_017009281.1 | Intron | XP_016864770.1 |