Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCAGCCTATACTGCAACTTTTT[A/G]TGCAAAGATTTTCAAATATTTTTTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613423 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCTD16 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KCTD16 - potassium channel tetramerization domain containing 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020768.3 | Intron | NP_065819.1 | ||||
XM_005268492.2 | Intron | XP_005268549.1 | ||||
XM_005268493.2 | Intron | XP_005268550.1 | ||||
XM_006714789.2 | Intron | XP_006714852.1 | ||||
XM_011537671.2 | Intron | XP_011535973.1 | ||||
XM_011537672.2 | Intron | XP_011535974.1 | ||||
XM_011537673.2 | Intron | XP_011535975.1 |
Set Membership: |
HapMap |