Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTTTACCATTACCACTCTTAAGA[C/T]GCTAGAAACAAGTAAGATTTAAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTAR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTAR1 - protein prenyltransferase alpha subunit repeat containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099666.1 | 8198 | UTR 3 | NP_001093136.1 | |||
XM_005251976.4 | 8198 | UTR 3 | XP_005252033.1 | |||
XM_005251977.4 | 8198 | UTR 3 | XP_005252034.1 | |||
XM_005251980.4 | 8198 | Intron | XP_005252037.1 | |||
XM_005251981.4 | 8198 | Intron | XP_005252038.1 | |||
XM_011518639.1 | 8198 | UTR 3 | XP_011516941.1 | |||
XM_011518640.1 | 8198 | UTR 3 | XP_011516942.1 |