Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTTATTGAGCCTTGAAGCTATCC[A/G]GTATTTATAAGAGGGATAAAATTAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609453 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GOLGA7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GOLGA7 - golgin A7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002296.1 | Intron | NP_001002296.1 | ||||
NM_001174124.1 | Intron | NP_001167595.1 | ||||
NM_016099.2 | Intron | NP_057183.2 |
Set Membership: |
HapMap |