Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAAAACCCAAAAACTGGATAGAG[C/T]CGAAAGGTACTTCAATGTGGTGGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PURG PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PURG - purine rich element binding protein G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015508.2 | 2095 | UTR 3 | NP_001015508.1 | |||
NM_001323311.1 | 2095 | Intron | NP_001310240.1 | |||
NM_001323312.1 | 2095 | UTR 3 | NP_001310241.1 | |||
NM_013357.2 | 2095 | Intron | NP_037489.1 |
Set Membership: |
HapMap |