Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAATGCTCAAAATTTTTGAACTA[A/T]GAATTCTGCCATTTTTCAGACAAGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605776 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FGL1 - fibrinogen like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004467.3 | Intron | NP_004458.3 | ||||
NM_147203.2 | Intron | NP_671736.2 | ||||
NM_201552.1 | Intron | NP_963846.1 | ||||
NM_201553.1 | Intron | NP_963847.1 | ||||
XM_017013232.1 | Intron | XP_016868721.1 | ||||
XM_017013233.1 | Intron | XP_016868722.1 |
Set Membership: |
HapMap |