Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCATTTTTGTCTTCTTGTGTAC[C/T]TCACATAATTTTTCTCCCTGATTTA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601487 | |||||||||||||||||||||||
Literature Links: |
PPARG PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PPARG - peroxisome proliferator activated receptor gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005037.5 | Intron | NP_005028.4 | ||||
NM_015869.4 | Intron | NP_056953.2 | ||||
NM_138711.3 | Intron | NP_619725.2 | ||||
NM_138712.3 | Intron | NP_619726.2 | ||||
XM_011533841.2 | Intron | XP_011532143.1 | ||||
XM_011533842.2 | Intron | XP_011532144.1 | ||||
XM_011533843.2 | Intron | XP_011532145.1 | ||||
XM_011533844.1 | Intron | XP_011532146.1 |