Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTCCTTTACCTCTTTACTCTCTC[C/T]TCAGACAAGCATTTACTGTGATGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610511 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEC23A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SEC23A - Sec23 homolog A, coat complex II component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006364.3 | Intron | NP_006355.2 | ||||
XM_005267262.1 | Intron | XP_005267319.1 | ||||
XM_011536355.2 | Intron | XP_011534657.1 | ||||
XM_017020928.1 | Intron | XP_016876417.1 |
Set Membership: |
HapMap |