Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCCACCAACCCAAGTGCTCTCTT[C/T]GCAGCTACGGTTCACAGTGAAGTCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
26 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601231 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105376736 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105376736 - uncharacterized LOC105376736 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MTOR - mechanistic target of rapamycin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004958.3 | Intron | NP_004949.1 | ||||
XM_005263438.2 | Intron | XP_005263495.1 | ||||
XM_011541166.2 | Intron | XP_011539468.1 | ||||
XM_017000900.1 | Intron | XP_016856389.1 | ||||
XM_017000901.1 | Intron | XP_016856390.1 | ||||
XM_017000902.1 | Intron | XP_016856391.1 |
Set Membership: |
HapMap |