Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTACTGAGGAAAGCTATGAGGATA[C/G]TCTGTTCGTAAGCTCCCGGTGAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616504 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929741 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101929741 - uncharacterized LOC101929741 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
METTL14 - methyltransferase like 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020961.3 | 48 | UTR 5 | NP_066012.1 | |||
XM_005263166.4 | 48 | UTR 5 | XP_005263223.1 | |||
XM_011532162.2 | 48 | UTR 5 | XP_011530464.1 |
Set Membership: |
HapMap |