Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCTTGGCTTGTACAATGGACCAC[A/T]GTCCCCAGAGGAAATGAGATGCAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 118491 MIM: 604592 | ||||||||||||||||||||
Literature Links: |
CHKA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHKA - choline kinase alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277.2 | 1363 | Missense Mutation | CAG,CTG | Q,L 422 | NP_001268.2 | |
NM_212469.1 | 1363 | Missense Mutation | CAG,CTG | Q,L 404 | NP_997634.1 | |
XM_017017147.1 | 1363 | Missense Mutation | CAG,CTG | Q,L 384 | XP_016872636.1 | |
XM_017017148.1 | 1363 | Missense Mutation | CAG,CTG | Q,L 246 | XP_016872637.1 |
TCIRG1 - T-cell immune regulator 1, ATPase H+ transporting V0 subunit a3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |