Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCACGGCCAGGTAGGCTGGCCC[G/C]CCTCTCTTCAGTCTGGTTGTCTGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604433 | ||||||||||||||||||||
Literature Links: |
KCNE3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCNE3 - potassium voltage-gated channel subfamily E regulatory subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005472.4 | 556 | Missense Mutation | CGG,GGG | R,G 47 | NP_005463.1 | |
XM_011544713.2 | 556 | Missense Mutation | CGG,GGG | R,G 91 | XP_011543015.1 | |
XM_017017047.1 | 556 | Missense Mutation | CGG,GGG | R,G 47 | XP_016872536.1 | |
XM_017017048.1 | 556 | Missense Mutation | CGG,GGG | R,G 47 | XP_016872537.1 | |
XM_017017049.1 | 556 | Missense Mutation | CGG,GGG | R,G 47 | XP_016872538.1 | |
XM_017017050.1 | 556 | Missense Mutation | CGG,GGG | R,G 47 | XP_016872539.1 | |
XM_017017051.1 | 556 | Missense Mutation | CGG,GGG | R,G 47 | XP_016872540.1 | |
XM_017017052.1 | 556 | Missense Mutation | CGG,GGG | R,G 47 | XP_016872541.1 |